Learn More
Edwards Fittings for BOC High Vacuum Pumps: Blank Flange
For use with BOC high vacuum pumps
15.90 €
Specifications
For Use With BOC high vacuum pump
Material Aluminum
View More Specs
Products 1
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
12925264
View Documents
Edwards
C10514368
NW25 Blank Flange
15.90 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
  • Made of aluminum
  • Aluminum is ideal for achieving dependable cost-effective performance down to 10-7mbar
Specifications
Specifications
BOC high vacuum pump
Aluminum