Learn More
Edwards Rotary Vane Vacuum Pump Fittings: Clamping Ring
Range of coupling clamps manufactured from high technology polymer for vacuum system builder
Brand:  Edwards C10514401
Product Code. 10462141
  • 16.40 € / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clamping ring: compact stainless-steel
  • Polymer clamp: lightweight, neat appearance
  • Aluminum clamp: rugged Pneurop standard
  • Metal clamp: suitable for aluminum and indium seals — wide temperature range
  • Claw clamps and bolts: wide range optimized for many applications — high strength CF bolts for UHV flanges
Specifications
Specifications
Rotary vane vacuum pump
Stainless Steel
Clamping Ring