Learn More
Scotsman™ EF124 Ice Flake Maker
The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access
Brand:  Scotsman™ EF124
Product Code. 12823226
  • 6850.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ice flakes are extruded at a temperature just below zero degrees and contain 20% residual water content. The flake ice is stored inside an insulated, plastic lined storage bin with front opening for easy access. Electrical supply 230V/50Hz.
  • Produces granular shape ice flakes, 80% hardness
  • Up to 120kg (2645 lbs) daily production
  • Up to 40kg (88 lbs) ice storage capacity
  • Ice scoop holder safely positioned in ice-storage bin
  • Resistant stainless steel exterior
  • Advanced diagnostics computerised control
  • Front panel in & out airflow (air-cooled model only) for built-in installation
  • Front access condenser air filter, removable and washable (air-cooled version only)
  • Easily accessible, front panel mount, ON-OFF switch
  • Easy, front panel access to technical / service area of the machine
  • 'Routine Maintenance' visible alarm light on front panel
  • Ergonomically designed ice storage access, with disappearing door
  • Door-closing movement dampening system
  • Vapour-based microbial control system pouch in ice storage area
  • Built -in PWD (pumped waste discharge) system for remote water drain (via easy-fit installation) for discharge of waste up to 1.5m above base or up to 20m from unit
  • AgION antimicrobial agent embedded in moulded plastics parts
  • CFC-free R-404A refrigerant
  • Two years manufacturer warranty on defective components
Scotsman flaked ice is ideal for a variety of applications within laboratory and industrial environments.
Specifications
Specifications
120 kg
50Hz
230V, 50Hz
Flaked Ice Maker
Grade 304 Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Scotsman™ EF124 Ice Flake Maker > includes waste pump, electronic controls, stainless steel finish 230V 50Hz 950mm x 605mm x 915mm (w x d x h) 120kg/24hrs max production, 40kg bin capacity Scotsman