missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EFHC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11471-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes an EF-hand-containing calcium binding protein. The encoded protein likely plays a role in calcium homeostasis. Mutations in this gene have been associated with susceptibility to juvenile myoclonic epilepsy and juvenile absence epilepsy. Alternatively spliced transcript variants have been described.Specifications
EFHC1 | |
Polyclonal | |
Unconjugated | |
EFHC1 | |
EFHC1, EJM, EJM1, JAE, Myoclonin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114327, 301295, 71877 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5JVL4, Q9D9T8 | |
Efhc1 | |
EFHC1 Fusion Protein Ag2048 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |