Learn More
EIF3A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26178-1-AP-150UL
 View more versions of this product
Product Code. 16894695
  • 410.00 € / 150µL
Explore more special offers
Description
Description
The German cDNA consortium.
Specifications
Specifications
EIF3A
Polyclonal
Unconjugated
EIF3A
eIF 3 theta, EIF3, eIF3 p167, eIF3 p170, eIF3 p180, eIF3 p185, eIF3 theta, EIF3A, EIF3S10, KIAA0139, P167, p180, p185, TIF32
Rabbit
Antigen Affinity Chromatography
RUO
8661
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.44 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q14152
EIF3A
EIF3A Fusion Protein Ag22515
150 μL