missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eighteen element solution for ICP/MS 10mg/L HNO3 matrix, Fisher chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
274.00 € - 516.00 €
Specifications
Chemical Name or Material | Eighteen Elements |
---|---|
Concentration | 10mg/L |
Packaging | Bottle |
Type | ICP/MS Calibration Multi-element Standard Solution |
Analyte or Component Names | Ag,Al,As,Ba,Be,Cd,Cr,Co,Cu,Mn,Ni,Pb,Se,Th,Tl,U,V,Zn |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12936193
|
Fisher Chemical
MS 008 050 |
50 mL | Bottle |
353.00 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12946193
|
Fisher Chemical
MS 008 100 |
100 mL | Bottle |
274.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12956193
|
Fisher Chemical
MS 008 250 |
250 mL | Bottle | N/A | |||||
Description
Eighteen elements solution - Ag, Al, As, Ba, Be, Cd, Cr, Co, Cu, Mn, Ni, Pb Se, Th, Tl, U, V, Zn for ICP/MS 10mg/L HNO3 matrixSpecifications
Eighteen Elements | |
Bottle | |
Ag,Al,As,Ba,Be,Cd,Cr,Co,Cu,Mn,Ni,Pb,Se,Th,Tl,U,V,Zn | |
Liquid |
10mg/L | |
ICP/MS Calibration Multi-element Standard Solution | |
2% HNO3 |
Safety and Handling
H412
P273;P501
missing translation for 'shelfLife' : 12 Months. (6 Months after opening)
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Multi-element Standard Solution for ICP. 18 Components in HNO3 2% (MS 008)
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .