Learn More
Invitrogen™ EK-Away™ Resin
Product Code. p-7182852
Click to view available options
Quantity:
7.5 mL
Unit Size:
Each
Product Code. 10403982
Brand: Invitrogen™ R18001
Includes
EK-Away is provided as a 50% ethanol slurry; 10X binding and 10X stripping buffers are included; 7.5mL resin will remove 250 units of enterokinase; 30mL will remove 1000 units of EKMax.
Specifically designed for removal of EKMaxor other enterokinase enzymes following cleavage of proteins containing the enterokinase cleavage site
  • The resin is conjugated with soybean trypsin inhibitor, which has a high affinity and binding capacity for enterokinase
  • The enzyme catalytic site binds this agarose-based resin for simple batch or column-bound removal of EKMaxor other enterokinase preparations
GST-Tagged Protein Purification, His-Tagged Protein Purification, Protein and Peptide Purification, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage EK-Away™ is provided as a 50% ethanol slurry and should be stored at +4°C. 10X binding and 10X stripping buffers are included. 7.5 ml resin will remove 250 units of enterokinase; 30 ml will remove 1000 units of EKMax™. All reagents are guaranteed stable for 6 months when stored properly.
Form Liquid Suspension
Product Line EK-Away
Quantity 7.5 mL
Type Resin
Stationary Phase Enterokinase
Column Type Affinity