missing translation for 'onlineSavingsMsg'
Learn More

Novex™ EK-Away™ Resin

EK-Away™ Resin

Brand:  Novex™ R18002

Product Code. 11573677

  • 2165.00 € / 30mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

EK-Away™ Resin is specifically designed for removal of EKMax™ or other enterokinase enzymes following cleavage of proteins containing the enterokinase cleavage site (Figure 1). The resin is conjugated with soybean trypsin inhibitor, which has a high affinity and binding capacity for enterokinase. The enzyme catalytic site binds this agarose-based resin for simple batch or column-bound removal of EKMax™ or other enterokinase preparations.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4°C
Resin
30 mL
Suspension
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novex™ EK-Away™ Resin > 30mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.