missing translation for 'onlineSavingsMsg'
Learn More

Manutan Ekwo three nest clean/dirty locker with feet, blue, cylinder lock, 1185mm wide

Product Code. 12847189
Encompass_Preferred
This item is not returnable. View return policy

Product Code. 12847189

Brand: Manutan A040339

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Strong single person locker with two vertical compartments and a hat shelf mounted on a plinth. Supplied in various colours, either mounted on four feet (for cleaning underneath) or on a plinth. Fitted with hat shelf, vertical central divider and a hanging rail in each compartment. Available with hasp lock (for use with a padlock - not supplied) or cylinder lock (supplied with two keys). Ventilation at top and bottom.

  • Three lockers supplied
  • Cylinder lock
  • Colour of body: Grey
  • Colour of door: Blue
  • Dimensions (H x W x D): 1900mm x 1185mm x 500mm
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.