missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Ekwo two nest clean/dirty locker with plinth, grey, cylinder lock, 800mm wide
Brand: Manutan A040357
This item is not returnable.
View return policy
Description
Strong single person locker with two vertical compartments and a hat shelf mounted on a plinth. Supplied in various colours, either mounted on four feet (for cleaning underneath) or on a plinth. Fitted with hat shelf, vertical central divider and a hanging rail in each compartment. Available with hasp lock (for use with a padlock - not supplied) or cylinder lock (supplied with two keys). Ventilation at top and bottom.- Two lockers supplied
- Cylinder lock
- Colour of body: Grey
- Colour of door: Grey
- Dimensions (H x W x D): 1800mm x 800mm x 500mm
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction