Learn More
Kolmi Hopen™ Elasticated Helmet Hood with Small Opening Product Code.: 17760139
Add to basket
Kolmi Hopen™ Elasticated Helmet Hood with Small Opening
Product Code. 17760139
Elasticated Helmet Hood with Small Opening; Color: White; Design Type: Helmet; Disposable: Yes; Includes: Caps and Polybags; Material: Polypropylene
Click to view available options
Elasticated Helmet Hood with Small Opening; Color: White; Design Type: Helmet; Disposable: Yes; Includes: Caps and Polybags; Material: Polypropylene
Product Code. 17760139
Brand: Kolmi Hopen™ 70.701
Add to basket
Add to basket
This elasticated helmet hood provides a high level of protection by leaving only the eyes exposed.
  • Ultra-effective protection for the wearer and the environment
  • Available for color code tracking
  • 25 g/m2 fabric weight
Specifications
Material Polypropylene
Color White
Disposable Single-use
Design Type Helmet
Quantity 50 Pk.
Includes Caps and Polybags
Product Content Correction
Product Title
Kolmi Hopen™ Elasticated Helmet Hood with Small Opening >