missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Electrode Saver Bottles

Keeps electrodes in working order with these electrode saver bottles. Cole Parmer™ Electrode Saver Bottles protect electrodes against bumps, spills and drying out. Accepts electrodes up to 13mm in diameter.

Brand:  Cole-Parmer™ 05990-99

35.73 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


 View more versions of this product

Product Code. 11795209

  • 40.35 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Keeps electrodes in working order with these electrode saver bottles. Cole ParmerTM Electrode Saver Bottles protect electrodes against bumps, spills and drying out. Accepts electrodes up to 13mm in diameter.

  • Large bottle includes one solid cap to reduce evaporation of storage solution and one O-ring cap for use with electrodes
  • Small bottle includes one O-ring cap and is a cost-effective option when storage solution is discarded and not regularly re-used
Specifications

Specifications

50 mL
Electrode Saver Bottle
One Solid Cap To Reduce Evaporation Of Storage Solution and One O-ring Cap For Use With Electrodes
279.4 mm
Electrodes
Protect Electrodes Against Bumps, Spills, and Drying Out
10 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Electrode Saver Bottles > Capacity: 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.