missing translation for 'onlineSavingsMsg'
Learn More

Mettler Toledo™ Electrolyte for Reference Electrodes 3 mol/L KCl Sat AgCl, 250mL

Mettler Toledo™ Electrolyte for Reference Electrodes 3 mol/L KCl Sat AgCl, 250mL can be used to fill your electrode with correct and fresh electrolyte for precise measurement.

Brand:  Mettler Toledo™ 51350074

24.29 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 10722712

  • 28.35 € / 250mL
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For use with pH and ORP electrodes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

3 mol/L KCl Sat AgCl
pH, ORP
250 mL
Electrolyte 3 mol/L KCl Sat AgCl, 250mL
250 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.