missing translation for 'onlineSavingsMsg'
Learn More

Bibby Scientific™ Electrothermal™ Heating Element for EM/EME

Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz

Brand:  Bibby Scientific™ 03012-85

 View more versions of this product

Product Code. 11768412

Quantity Price
1 585.00 € / Each
5 439.34 € / Each
Estimated Shipment: 15-05-2025
to see stock.
Explore more special offers
Add to basket

This item is not returnable. View return policy

Description

Description

Bibby Scientific Electrothermal Heating Element for EM/EME, Capacity: 250 mL, For Use With: 14314 EM Series Heating Mantles, Voltage: 230 V, Frequency: 50/60 Hz

Specifications

Specifications

5000 mL
230 V
Heating Element
EM/EME Series Heating Mantles
800w
50/60 Hz
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bibby Scientific™ Electrothermal™ Heating Element for EM/EME > Capacity: 5000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.