missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ elements™ Modular Stirrer Control Unit

Completely customizable stirrer designed to optimize your stirring needs and workspace.

Brand:  Cole-Parmer™ 84003-81

Product Code. 16416629

  • 203.00 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Attach up to 10 stir plates based on need
  • Simple touch and slide operation
  • LCD display with real time parameters
  • Corrosion-resistant glass stir top
  • Waterproof USB attachments
  • Specifications

    Specifications

    Up to 10 L
    Modular Stir Plates
    50/60 Hz
    100 to 1500 rpm
    Control Unit
    Controls up to 10 modular units
    Multi-position Stirrer
    Control module, power cord, and AC adapter
    5 to 40°C
    Digital LCD
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.