Learn More
Tocris Bioscience™ Elinogrel
P2Y12 antagonist
Brand:  Tocris Bioscience™ 5316/10
188.34 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18766248
  • 213.00 € / 10mg
Explore more special offers
Description
Description
Elinogrel is a P2Y12 antagonist; exhibits antiplatelet activity.
Specifications
Specifications
97%
10 mg
523.95
-20°C
Elinogrel
Elinogrel