Learn More
Bel-Art™ SP Scienceware™ Ellipso-Spoon™ Spatula
Specially curved shape permits contact with all corners of bottles and containers to minimize sample loss
Brand:  Bel-Art™ SP Scienceware™ H36806-0018
Product Code. 15590122
  • 33.13 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Extracts crystals, granulated and solid substances, thick liquids, and delicate materials
  • Features a spoon end and a spatula end
  • Made of polished 304 grade stainless steel
  • Flameproof, autoclavable, and impervious to boiling water
Specifications
Specifications
Spoon and Spatula Sampler
Autoclavable
Stainless Steel
1
15 mm
Stainless Steel
10 mL
None
18 cm
35 mm