Learn More
Bel-Art™ SP Scienceware™ Ellipso-Spoon and Spatula Samplers
Special curved shape prevents missed sample accumulation in corners.
Brand:  Bel-Art™ H36806-0015
 View more versions of this product
Product Code. 12907320
  • 36.75 € / Each
Estimated Shipment: 21-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Crystals, powders, granulated and solid substances, and thick liquids retrieved
  • Includes spoon end and spatula end
  • Made of polished 304 grade stainless steel
Specifications
Specifications
Spoon and Spatula Sampler
Autoclavable
15 x 35 mm bowl
Designed to extracts crystals, granulated and solid substances, thick liquids and delicate materials
15 cm
Stainless Steel
10 mL Bowl
Polished
Bottles and containers