missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Ellipso-Spoon and Spatula Samplers

Special curved shape prevents missed sample accumulation in corners.

Brand:  Bel-Art™ H36809-0050

 View more versions of this product

Product Code. 12917320

  • 101.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Crystals, powders, granulated and solid substances, and thick liquids retrieved
  • Includes spoon end and spatula end
  • Made of polished 304 grade stainless steel
Specifications

Specifications

Spoon and Spatula Sampler
Autoclavable
70 mL bowl
Curved
8 mm dia.
Stainless Steel (AISI 304)
Spoon end and spatula end
28 x 65 mm bowl
50 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bel-Art™ Ellipso-Spoon and Spatula Samplers > Bowl Capacity: 70mL; Bowl size: 28 x 65mm; Handle dia. 8mm; 50cm L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.