missing translation for 'onlineSavingsMsg'
Learn More

Eluent Concentrate, PRP-X100 Anion Exchange, Hamilton™

Hamilton chromatography resins can be ordered in bulk, i.e. not packed into column hardware. This is a low cost approach to application-specific testing and analyses.

Brand:  Hamilton 79335


Product Code. 11537904

  • 831.00 € / Pack of 6

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specification Sheet

Separation ModeAnion Exchange
FormatEluent Concentrate
Sales Quantity6 x 60 mL bottles
Composition4.0 mM p-Hydroxybenzoic Acid / 2.5% Methanol
*$75 will be added to shipping for DOT hazardous material classification*
Specifications

Specifications

4.0 mM p-Hydroxybenzoic Acid / 2.5% Methanol
6 x 60 mL bottles
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • EC-X100 eluent concentrate

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Acute toxicity Category 4

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.