Learn More
Invitrogen™ Silencer™ Select Human Drug Targets siRNA Library (384 well)
The Silencer Select Human Drug Targets siRNA Library contains 23,778 Invitrogen™ Silencer Select highly potent and chemically-modified siRNAs targeting 7,926 genes for performing RNAi screens against known human drug target genes.
Brand:  Invitrogen™ A30091
Product Code. 15601862
  • 107000.00 € / 1 set
Explore more special offers
Description
Description
The Silencer™ Select Human Drug Targets siRNA Library includes genes involved in apoptosis, senescence, nucleic acid binding, autophagy, DNA repair, along with characterized nuclear receptors and other genes that are considered potential targets for therapeutics. The Silencer Select Human Drug Targets siRNA Library contains 23,778 Ambion™ Silencer Select highly potent and chemically-modified siRNAs targeting 7,926 genes for performing RNAi screens against known human drug target genes. This library was designed using the most up-to-date genome databases, including the NCBI RefSeq database and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC).
Why use the Silencer Select Human Drug Targets siRNA Library?Together with careful bioinformatics selection of gene targets, Silencer Select siRNAs are optimized with latest design algorithms, proprietary chemical modifications and high quality synthesis to help ensure desired RNAi outcomes. The Silencer Select Human Drug Targets siRNA Library contains our pre-designed and when available, validated siRNAs generated with highest potency and specificity in mind so you can focus on the highest data quality backed by the best in industry guarantee.Silencer Select siRNA libraries empower you with highly potent siRNAs to maximize discovery with even lessInvitrogen Silencer Select siRNAs are designed with the highest potency of available siRNAs, resulting in 100-fold more potent siRNAs and allowing gene knockdowns at the lowest concentrations.The highly potent Silencer Select siRNAs allow you to use even lower concentrations of siRNAs, which helps to minimize off target effects, reduce false negative data and maximize siRNA library output for additional screens and data validation. The Silencer Select Human Drug Targets siRNA Library contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume).
The library includes gene IDs from Thermo Fisher’s Human Druggable Genome and Human Extended Druggable Genome siRNA libraries that are not already in the kinase, phosphatase, protease, ion channel, GPCR or ubiquitin libraries. The Silencer Select siRNAs are designed using Thermo Fisher Scientific’s leading algorithms—along with our novel locked nucleic acid (LNA) chemical modifications, this helps to help ensure the efficacy, specificity and potency required for RNAi screening using a comprehensive pre-defined platform for therapeutic discovery.
  • Superior efficacy —Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
  • Highest efficiency—Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
  • Dependable—Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
  • Convenient—Reduce data de-convolution and accelerate target validation with our 96- and 384-well arrayed format
  • Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats
TRUSTED_SUSTAINABILITY
Specifications
Specifications
23,778 total siRNAs targeting 7926 genes plated onto 384-well plates (69 plates)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For research use only. Not for use in diagnostic procedures