missing translation for 'onlineSavingsMsg'
Learn More

Merck Chemical Duty Vacuum Pressure Pump

Decrease process filtration times. Chemical Duty Vacuum Pressure Pump supports high flow rates for faster pumping. Cast-aluminum pumphead, housing, and regulator with optional chemical-resistant internal components. Light weight and portable so they can be shared among workstations.

Brand:  Merck WP6111560

 View more versions of this product

Product Code. 10555950

  • 1775.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket

Alert:

220v model is a non-standard voltage; please ensure you are ordering the correct product.


Includes: 70cm of .25 in. dia. tubing; Millex™ FA50 filter for in-line moisture protection.
This item is not returnable. View return policy

Description

Description

Decrease process filtration times. Chemical Duty Vacuum Pressure Pump supports high flow rates for faster pumping. Cast-aluminum pumphead, housing, and regulator with optional chemical-resistant internal components. Light weight and portable so they can be shared among workstations.

  • High output pumps feature a piston-driven design for greater power and high flow rates to decrease process filtration time
  • With optional chemical resistant head and diaphragm for use with corrosive chemicals, solvents and vapors
  • Portable design allows for easy sharing between workstations
  • Include a Millex™ FA50 filter for in-line moisture protection
  • UL listed and CE marked

Certifications

UL listed and CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

813 mbar (24 in.Hg)
4.1 kg
115 V 60 Hz
9 lb.
37 L/min.
7 in.
8 in.
2.45 bar
9 lb.
70cm of 0.25 in. dia. tubing and Millex™ FA50 filter
Aluminum
4.1 kg
60 Hz
17.8 cm
20.3 cm
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers