missing translation for 'onlineSavingsMsg'
Learn More

Merck Empty Monitor Cassette for 37mm Aerosol Analysis Monitors

For 37mm Monitor

Brand:  Merck M000037A0

 View more versions of this product

Product Code. 10577872

  • 519.00 € / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Empty aerosol analysis monitors
  • For use with Type MF (mixed cellulose esters) Millipore filters

Millipore's 37 mm Aerosol Analysis Monitors are transparent, disposable, plastic filter holders for use with Type MF (mixed cellulose esters) Millipore filters. A thin cellulose support seals the filter between monitor sections to distribute sample flow evenly over the filter surface.

A three-part design holds the filter in-place when the top section is removed for open aerosol sampling. The Average Background particle Count (ABC) value is marked on each monitor package.

Applications:
Aerosol analysis monitors are available with 0.45 μm and 0.8 μm cellulose esters membranes. Matched weight monitors contain two filters matched in weight to within 0.1 mg, eliminating test filter pre-weighing in gravimetric analysis.

Order Info

Bulk pack.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aerosol Analysis Monitor Case
Female Luer Slip
To reload cassette with appropriate filters
Polystyrene
EMD Millipore Aerosol Analysis Monitor (37 mm)
Empty monitor cassette with rings and plugs
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.