Learn More
Merck Immobilon™-P PVDF Membrane
Hydrophobic PVDF Transfer Membrane for western blotting
278.00 € - 941.00 €
Specifications
Material PVDF
Pore Size 0.45 μm
View More Specs
Products 10
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
Product Code Brand Length (Metric) Width (Metric) Format Price Quantity & Availability  
10408793
View Documents Promotion Details
Merck
IPVH08100
8 cm 10 cm Sheet
278.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10344661
View Documents Promotion Details
Merck
IPVH00010
26.5 cm 3.75 m Roll
822.00 €
1 roll
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
10503224
View Documents Promotion Details
Merck
IPVH08130
8.5 cm 13.5 cm Sheet
346.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10505672
View Documents Promotion Details
Merck
IPVH07850
7 cm 8.4 cm Sheet
877.00 €
Pack of 50
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10458712
View Documents Promotion Details
Merck
IPVH09120
9 cm 12 cm Sheet
278.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10663751
View Documents Promotion Details
Merck
IPVH10100
10 cm 10 cm Sheet
340.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10384511
View Documents Promotion Details
Merck
IPVH15150
15 cm 15 cm Sheet
425.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10180771
View Documents Promotion Details
Merck
IPVH20200
20 cm 20 cm Sheet
618.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
10796851
View Documents Promotion Details
Merck
IPVH304F0
26 cm 26 cm Sheet
941.00 €
Pack of 10
Estimated Shipment: 26-11-2024
to see stock.
Add to basket
16329601
View Documents Promotion Details
Merck
IPVH85R
10 m 8.5 cm Roll
742.00 €
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
Description
Description
Immobilon™ PVDF membranes are offered in four types, each optimized for a different protein blotting application. Convenient blotting sandwiches feature pre-cut sheets of membrane and blotting filter paper. Immobilon™ PVDF membranes have high protein adsorption, so you won’t lose proteins during transfer or reprobing. The open pore structure makes it easy to access bound proteins and remove unbound probes. In addition, Immobilon™ PVDF membranes optimized for fluorescent blots dramatically increase signal-to-noise ratios for high sensitivity in quantitative, multiplexing applications.
Specifications
Specifications
PVDF
0.45 μm