missing translation for 'onlineSavingsMsg'
Learn More

Merck Millipore Microfil™ Funnels

Available in different colors


Description

Description

Available in different colors

  • 0.22μm pore size for specific test regulations
  • 0.45μm for general microbiological analysis
  • 0.7μm for chlorinated effluents
  • 0.8μm for yeast or molds applications

Order Info

47mm gridded MCE membrane filters or EZ-Pak™ membrane cartridge

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Kit: 150xMicrofil 250ml funnel + 150x0.45µm 47mm white gridded EZ-Pak membrane (cellulose esters)

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P370+P378-In case of fire: Use powder to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : nitrocellulose

SDS
Documents

Documents

Product Certifications