Learn More
Merck Millipore Milli-Q™ Ultrapure Water Systems Accessory, Cartridges
Choose from a wide selection of replacement cartridges
Brand:  Merck Millipore QGARD00R1
 View more versions of this product
Product Code. 12949343
  • 987.00 € / Each
Explore more special offers
Description
Description
Choose from a wide selection of replacement cartridges
Filter Types
  • Activated carbon
  • Ion exchange
  • Membrane