Learn More
Merck Millicell™ Culture Plate Inserts
Designed for improved cell differentiation and expanded applications involving analytical cell biology. Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.
Brand: Merck PICM01250
Description
Designed for improved cell differentiation and expanded applications involving analytical cell biology. Millicell™ Culture Plate Inserts incorporate PTFE Biopore™ membranes with 0.4μm pores. Ready-to-use 30 or 12mm inserts for 6 and 24-well plates.
Microporous, transparent membranes- Allow access to cell monolayer from both apical and basolateral domains
- Promote enhanced cell attachment, growth, and differentiation of attachment-dependent and suspension cultures
- Hydrophilic PTFE Biopore™ membranes for neural growth and embryo development work
- Inert and allow repeated viewing of living cells through the membrane
- Suitable for transport and permeability studies, co-culture for cell-cell interaction studies, in vitro toxicity testing, tumor cell metastasis studies, and organotypic cultures
- Fit most 6- or 24-well plates and are easily prepared for SEM and TEM techniques
- Effective growth area for 12mm diameter inserts: 0.6cm2
- Effective growth area for 30mm diameter inserts: 4.2cm2
- Compatible with standard and fluorescent stains
Specifications
0.6 cm2 | |
0.4 μm | |
24 Well Plate | |
10.5mm | |
Sterile |
Biopore PTFE | |
Culture Insert | |
12mm | |
6-, 24-Well Plates | |
6, 24 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.