Learn More
Merck Millicell-ERS2 Volt-Ohm Meter and accessories
Measures Trans Epithelial Electrical Resistance (TEER) of epithelial cells in culture
Brand:  Merck MERSSTX03
 View more versions of this product
Product Code. 11750363
  • 1075.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Includes: Millicell-ERS2 comes with meter, battery, STX01 electrode, 1,000 Ω test electrode, battery charger. Specialized electrodes for Millicell-96 well plate are available (Cat. No. MERSSTX00).
Description
Description
Provides qualitative measurement of cell monolayer health and quantative measures of confluency. Designed exclusively for use with cells cultured in Millicell™ Culture Plate Inserts.
Utilizes AC current for membrane resistance measurements, producing a zero net charge on cells and eliminating adverse DC current effects on cell membranes. A silver/silver chloride (Ag/AgCI) pellet on each electrode tip measures voltage. Due to the small size of the electrodes, the user can easily measure transepithelial voltage and the resistance of cells grown on microporous membranes.
Services
Electrodes and batteries are warranted for 30 days only from the date on which the customer receives these items
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adjustable Electrode
19 cm
2.3 in.
12.5 Hz
Adjustable Electrodes, 1 pair
8 to 10 hr.
10° to 38°C
1.4 kg
7.3 in.
To provide qualitative measurement of cell monolayer health and quantative measures of confluency
6 cm
0 to 90%
1 Pair
50° to 100°F
3 lb.
11 cm