Learn More
Merck Millicell™ EZ Slides
Culture, fix, stain, and view cells all in one device
Brand:  Merck PEZGS0896
 View more versions of this product
Product Code. 11705264
  • 1495.00 € / Pack of 96
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polypropylene wells are resistant to most fixatives—cells may be fixed and stained with wells still attached
  • Breakable tabs for easy well removal—no tools required to remove wells from slide
  • No adhesive used to attach wells to slide
  • No leaky wells
  • Raised corner ridges on lid facilitate stacking of multiple units in the incubator
  • Noncytotoxic and nonpyrogenic
  • Sterile
  • Slide holder (available separately) protects slide from damage
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EZ Slide
Clear
0.7 cm2/well
Glass
Simplify your cell analysis by using the Millicell EZ SLIDE glass slide to culture, fix, stain and view your sample all in one device
8
Polypropylene
Polystyrene
0.2 to 0.7 mL/Well
Sterile