Learn More
Merck MultiScreen™ 96-Well Assay Plates for Sample Preparation
For the isolation, purification, and preparation of samples prior to testing in drug discovery processes and protein and DNA applications
534.00 € - 2695.00 €
Specifications
Format Standard
Array 8 x 12
No. of Wells 96
View More Specs
Products 6
Product Code Brand Color Material Sterility Pore Size Price Quantity & Availability  
Product Code Brand Color Material Sterility Pore Size Price Quantity & Availability  
10470211
View accessory products View Documents
Merck
MABVN1250
Clear Styrene Non-sterile 1.2 μm
2465.00 €
Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
10370031
View accessory products View Documents
Merck
MADVN6550
Clear Styrene Non-sterile 0.65 μm
2465.00 €
Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
10607891
View Documents
Merck
MAGVS2210
Clear ABS Plastic Sterile 0.22 μm
687.00 €
Pack of 10
Estimated Shipment: 04-03-2025
to see stock.
Add to basket
10711801
View accessory products View Documents
Merck
MAHVN4510
Clear Styrene Non-sterile 0.45 μm
534.00 €
Pack of 10
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
10054540
View accessory products View Documents
Merck
MAIPN0B50
Opaque Barex Non-sterile 0.45 μm
2695.00 €
Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
10658271
View accessory products View Documents
Merck
MAHVN4550
Clear Styrene Non-sterile 0.45 μm
2545.00 €
Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Description
Description
Formats are suitable for removal of large particulates, resin-based separations, solvent applications, and protein/DNA concentration or purification. Versatile separation system combines a microplate with Millipore's membrane filtration to simplify and optimize drug discovery techniques, biochemical assays, and traditional sample preparation.
  • Use with a vacuum manifold, automated liquid-handling system, or microplate scintillation counter—entire assays can be performed on one plate
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
  • Also ensures quantitative filtrate transfer to a receiver plate placed in bottom of manifold
Choice of filter materials
  • For high protein binding applications, use plates with Immobilon™-P PVDF or mixed cellulose esters For high nucleic acid binding, use plates with mixed cellulose esters, glass fiber, or positively charged DEAE membranes For charged biomolecule separations, choose from positively charged DEAE or negatively charged phosphocellulose membranes
  • For low biomolecule binding, use plates with hydrophilic Durapore™ PVDF, Biopore™ PTFE, or polycarbonate membranes
  • For assays that require solvent resistance, use plates with hydrophilic PTFE membranes
Each microporous membrane interacts distinctly with different proteins or biomolecules; therefore, it may be necessary to test different membrane types for each assay.
Specifications
Specifications
Standard
8 x 12
96