Learn More
Merck MultiScreen™ Carrier Racks
For radionucleotide counting by transfer of individual filters to tubes
Brand:  Merck MACR81275
 View more versions of this product
Product Code. 10507492
  • 835.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Carrier racks hold sample collection containers in place; available for 4mL vials, 7mL vials, or 12 x 75mm tubes (12 racks per carrier rack assembly)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Carrier rack for 12 x 75mm tubes
Radionucleotide counting by transfer of individual filters to tubes
1/Pk.