Learn More
Merck MultiScreen™ Centrifuge Alignment Frame
Allows easy and accurate transfer of samples
Brand:  Merck MACF09604
Product Code. 10616551
  • 118.00 € / Pack of 4
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Transfer samples filtrate to the wells of a 96-well collection plate
  • Prevents spillage or cross-contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Blue
Non-sterile
4/Pk.
ABS Plastic
Use to align MultiScreen plate to 96-well receiver plate for centrifugation of aqueous media