missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™ Column Loaders
Shop All Merck Products
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
Unit Size:
Each
Description
- Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Color | Black |
| Volume (Metric) Well | 25 μL |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| For Use With (Application) | Load 25μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography |
| Quantity | 1/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction