Learn More
Merck MultiScreen™ Multiple Punch Kits
Includes punch with punch carrier plate and punch distributor
Brand: Merck MAPK8960A
This item is not returnable.
View return policy
Description
Specifications
Radionucleotide counting by transfer of individual filters to tubes | |
Multiple punch, 2 carrier racks for 7mL vials | |
1/Pk. |