Learn More
Merck MultiScreen™ Multiple Punch and Punch Tips
For rdionucleotide counting by transfer of individual filters to tubes
677.00 € - 7125.00 €
Specifications
For Use With (Application) Radionucleotide counting by transfer of individual filters to tubes
View More Specs
Products 2
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
10156100
View Documents
Merck
MADP19650
Punch tip
677.00 €
Pack of 50
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
11546365
View Documents
Merck
MAMP09608
Punch kit
7125.00 €
Pack of 10
Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Description
Description
  • Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
  • Remove filters from wells separately, eliminating cross-contamination
  • Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
  • Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifications
Specifications
Radionucleotide counting by transfer of individual filters to tubes