missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane
8-well strips enable processing of less than 96 samples at one time
278.00 € - 1630.00 €
Specifications
Format | Strip Plate |
---|---|
Array | 1 x 8 |
Color | Clear |
Material | ABS Plastic |
No. of Wells | 8 |
Includes: Twelve 8-well strips in a support frame. Additional support frames also available separately.
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
10169794
|
Merck
M8IPS4510 |
Microplate 8-Well Strip |
1630.00 €
Pack of 10 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
10174073
|
Merck
M8IPFRAME |
Microplate Strip Support Frame |
278.00 €
Pack of 10 |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||||
Description
- For ELISpot and protein binding assays
Specifications
Strip Plate | |
Clear | |
8 |
1 x 8 | |
ABS Plastic | |
Sterile |