Learn More
Merck MultiScreen™HTS 96-Well Filter Plates for Clearing
Designed to meet specific performance criteria for sample preparation methodology
Brand: Merck MSNANLY10
This item is not returnable.
View return policy
Description
- Automation-compatible multiscreen filter plates
- Lysate clearing
Clearing of bacterial cell lysates
Specifications
Standard | |
Clear | |
96 | |
Clearing of bacterial cell lysates |
8 x 12 | |
Styrene | |
Non-sterile | |
10 |