missing translation for 'onlineSavingsMsg'
Learn More

Merck Pellicon™ XL Devices

Product Code. 10184739 Shop All Merck Products
Click to view available options
MWCO:
10 kDa
30 kDa
5 kDa
50 kDa
pH Range:
1 to 14
2 to 21
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10184739

Brand: Merck PXB010A50

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Operating instructions and Certificate of Quality

Small-volume tangential flow filter device

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.

Order Info

Packaged one per box.

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Cassette
Material Polypropylene
MWCO 10 kDa
Pore Size 100 μm
Filtration Area 50 cm2
For Use With (Application) Protein purification, analytical sample preparation
Includes Operating instructions and certificate of quality
Max. Inlet Pressure 80 psi at 50°C
pH Range 1 to 14
Quantity 1 Pack
Recirculation Rate 0.03 to 0.05L/min.
Type Pellicon XL Ultrafiltration Device
Volume (English) Working 0.5 oz.
Volume (Metric) Hold Up 3.2 mL
Volume (Metric) Working 15 mL
Dimensions (L x W) 7.4 x 1.2 in. (18.8 x 3cm)
Length (English) 7.4 in.
Length (Metric) 18.8 cm
Width (English) 1.2 in.
Width (Metric) 3 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.