missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck PureProteome™ Carboxy FlexiBind Magnetic Bead System
For rapid and reproducible isolation of proteins
880.00 € - 2375.00 €
Specifications
Product Type | Carboxy flexiBind Magnetic bead system |
---|---|
Content And Storage | 2° to 8°C |
For Use With (Application) | Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions |
Matrix | Magnetic Beads |
Product Code | Brand | Quantity | Particle Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Particle Size | Price | Quantity & Availability | ||||
12691146
|
Merck
LSKMAG1CBX10 |
10mL | 1μm |
880.00 €
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
12601156
|
Merck
LSKMAG25CBX10 |
10mL | 2.5μm |
2375.00 €
Each |
Estimated Shipment: 28-11-2024
Log in to see stock. |
||||
Description
Custom affinity bead generation for binding to target ligand.
- Advanced chemistry combined with high surface area provides more binding sites for proteins
- Particles visible as they adhere to side of tube for quick and easy aspiration and complete buffer removal
- Beads are immobilized in seconds
- Increased kinetics of bead-protein binding enables shorter incubations
- Significantly more affordable then competitive magnetic beads
Research Use Only
Specifications
Carboxy flexiBind Magnetic bead system | |
2° to 8°C | |
Immunoprecipitations, purifying Nucleic Acids, isolating cells/organelles, Protein-protein interactions | |
Magnetic Beads |