Learn More
Merck PureProteome™ Nickel Magnetic Beads
Offer yields comparable with other magnetic purification systems while offering high purities
Brand:  Merck LSKMAGH02
 View more versions of this product
Product Code. 11730353
  • 308.00 € / Pack of 2
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • No lysate clearing—crude cell lysates are processed by magnetic beads
  • Fast processing time—entire protocol is completed in less than one hour
  • High capacity—bind 5 to 15mg of protein per mL of settled beads
  • High affinity—binding can take place in buffer containing <5mM EDTA
  • Flexible scale—varying the bead quantity allows different protein yields
  • Flexible elution—native or denatured proteins
  • Automatable—hands-free screening process
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 to 5.5mg/mL
10 μm
Purification of His-tagged recombinant Protein
2°C to 8°C
2 x 1 mL
Magnetic Affinity Bead