Learn More
Merck RC800 Mini-Reservoir
Accessory for simplified stirred cell operation
Brand:  Merck 6028
Product Code. 10783291
  • 3470.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Graduated acrylic body
  • Acetal cover, removable for easy cleaning
  • Acetal fittings, Buna-N, silicone O-rings
  • Safety valve for slow pressure bleed (max. pressure: 5 bar)
  • Polyethylene tubing
  • Wrap-around clamp for extra safety
  • Chemically sanitizable