Learn More
Merck Samplicity™ Filtration System Vial Trays
Vacuum filtration system for sample preparation for HPLC
Brand:  Merck SAMWASTTR
 View more versions of this product
Product Code. 12541249
  • 131.00 € / Pack of 5
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Overview of Samplicity™ G2 System
  • Samplicity™ G2 Filtration System enables vacuum driven filtration of up to 8 samples at once, directly into HPLC vials.
  • Filtration for HPLC sample prep without syringes. Sample is added to an adapter funnel on a standard Millex™ 33 mm filter (Millex™ Filters for the Samplicity™ G2 System) or an integrated funnel and filter (Millex Samplicity™ filter)
  • Samplicity™ G2 System has lower ergonomic impact than manual syringe filtration
Applications: HPLC; Filtration of viscous or particulate laden samples for HPLC
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Samplicity G2 Filtration System
Non-sterile
5 Trays
Recycled Polyester
Analytical Sample Prep, High Performance Liquid Chromatography (HPLC)
Waste Trays