Learn More
Merck Accessories and Replacement Parts In-Line Stainless Steel Filter Holders 47 mm
Accessories for filter holders for sample preparation.
Brand:  Merck XX4204709
 View more versions of this product
Product Code. 10460103
  • 104.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
XX4204709, XX4304701, XX4404703, 5614, and 00907P are replacement parts for Filter Holder XX4404700
SS Filter, Depends on filter type used.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Not Autoclavable
47 mm high Pressure SS filter holder
Non-sterile
Stirred Cell Support Screen
Stainless Steel
Support Screen, 47 mm dia. Filter, Stainless Steel