missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Accessories for ZHE Hazardous Waste System
Accessories for filter holders for sample preparation.
Brand: Merck YY2209064
This item is not returnable.
View return policy
Description
Specifications
Not Autoclavable | |
ZHE Hazardous Waste filtration system | |
Non-sterile | |
90 mm |
Support Screen 90mm | |
Stainless Steel | |
Support Screen 90 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction