missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Emtricitabine Product Code.: 15781789

Thermo Scientific Chemicals Emtricitabine

Product Code. 15781789
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15781789

Brand: Thermo Scientific Chemicals 462070010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143491-57-0
Molecular Formula C8H10FN3O3S

Specifications

Quantity 1 g
Solubility Solubility in water: soluble. Other solubilities: very soluble in methanol,very slightly soluble in acetonitrile
Merck Index 153622
Melting Point 136°C to 140°C
Formula Weight 247.24
Chemical Name or Material Emtricitabine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0446130 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog Number462070050,462070010
Lot NumberA0444785 Certificate TypeCertificate of Analysis Date08/02/2024 Catalog Number462070050,462070010
Lot NumberA0442005 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog Number462070050,462070010
Lot NumberA0436496 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number462070050,462070010
Lot NumberA0439315 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number462070050,462070010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Emtricitabine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.