missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Enamelled Steel Container

Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.

Brand:  Day Impex™ 200013

 View more versions of this product

Product Code. 11786399

  • 343.00 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Safe for use with all liquid gases and ideal for cold traps
  • Attractive blue stove-enamelled steel outer container
Specifications

Specifications

182mm
280mm
With lid and handle
280 mm
Enameled Steel
Use for liquid gas or as a cold trap
350mm
4500 mL
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers