missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Enamelled Steel Container

Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.

Brand:  Day Impex™ 200004

 View more versions of this product

Product Code. 11827452

  • 208.00 € / Each
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Safe for use with all liquid gases and ideal for cold traps
  • Attractive blue stove-enamelled steel outer container
Specifications

Specifications

81mm
315mm
Without handle or lid
315 mm
Enameled Steel
Use for liquid gas or as a cold trap
350mm
700 mL
54 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.