missing translation for 'onlineSavingsMsg'
Learn More

Day Impex™ Enamelled Steel Container

Safely store and transport liquid gases. Day Impex™ Enamelled Steel Container is specially designed for laboratory, hospital, and industrial use.

Brand:  Day Impex™ 200005

 View more versions of this product

Product Code. 11837452

  • 201.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Safe for use with all liquid gases and ideal for cold traps
  • Attractive blue stove-enamelled steel outer container
Specifications

Specifications

94mm
240mm
Without handle or lid
240 mm
Enameled Steel
Use for liquid gas or as a cold trap
285mm
750 mL
65 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers