missing translation for 'onlineSavingsMsg'
Learn More
Learn More
endo-IWR 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
endo-IWR 1 is an inhibitor of Wnt signaling. Induces an increase in axin2 protein levels; promotes β-catenin phosphorylation by stabilizing Axin-scaffolded destruction complexes. Promotes endothelial cell specification of cardiac progenitor cells
Negative Control also available.
For more information about how endo-IWR 1 may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 409.44 |
Percent Purity | >98% |
Chemical Name or Material | endo-IWR 1 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'