missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Endothall
Click to view available options
Quantity:
20 mg
100 mg
Description
Endothall is used as an inhibitor of PP2A phosphatase activity. It is also used as a defoliant. It is used as a cell adhesion inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Quantity | 100 mg |
Description | White |
Content And Storage | 2°C to 8°C |
For Use With (Application) | A moderately potent inhibitor of protein phosphatase 2A |
Formula Weight | 186.20g/mol |
Product Type | Signal Transduction Reagent |
Form | Powder |
InChI Key | XRHVZWWRFMCBAZ-UHFFFAOYNA-L |
SMILES | [Na+].[Na+].[O-]C(=O)C1C2CCC(O2)C1C([O-])=O |
IUPAC Name | disodium 7-oxabicyclo[2.2.1]heptane-2,3-dicarboxylate |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction